Ethyl 2-ethoxy-4-methylbenzoate 97%

Supplier: Ambeed
Warning

A211669-1G A211669-250MG
76825-920EA 66.84 USD
76825-920 76825-922
Ethyl 2-ethoxy-4-methylbenzoate 97%
Ethyl 2-ethoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.25 g/mol
MDL Number: MFCD09701458
CAS Number: 88709-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR