Diethyl 2-(1-(4-acetamidophenyl)-1-oxopropan-2-yl)malonate 98%

Supplier: Ambeed
Warning

A211463-1G A211463-250MG
76825-916EA 338 USD
76825-916 76825-918
Diethyl 2-(1-(4-acetamidophenyl)-1-oxopropan-2-yl)malonate 98%
Diethyl 2-(1-(4-acetamidophenyl)-1-oxopropan-2-yl)malonate

Precaución: for Research Use Only

Formula: C₁₈H₂₃NO₆
MDL Number: MFCD23135508
CAS Number: 81937-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR