Methyl 5-bromo-2-formylbenzoate 95%

Supplier: Ambeed
Warning

A211023-100MG A211023-250MG A211023-10G A211023-5G
76825-904EA 25.47 USD
76825-904 76825-906 77593-378 77612-080
Methyl 5-bromo-2-formylbenzoate 95%
Methyl 5-bromo-2-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.054 g/mol
MDL Number: MFCD16036575
CAS Number: 1016163-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR