Cinnamyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

A210953-1000G A210953-100G A210953-25G A210953-500G
76825-896EA 47 USD
76825-896 76825-898 76825-900 76825-902
Cinnamyl alcohol 95%
Cinnamyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₀O
MDL Number: MFCD00002921
CAS Number: 104-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR