6-Fluoro-4-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A210770-100MG A210770-250MG A210770-1G
76825-892EA 10 USD
76825-892 76825-894 77885-516
6-Fluoro-4-nitro-1H-indazole 97%
6-Fluoro-4-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄FN₃O₂
MDL Number: MFCD07781404
CAS Number: 885520-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR