5-Bromo-4-fluoropyridin-2-amine 98%

Supplier: Ambeed
Warning

A210698-100MG A210698-1G A210698-250MG A210698-25G A210698-5G
76825-884EA 6 USD
76825-884 76825-886 76825-888 76825-890 77568-036
5-Bromo-4-fluoropyridin-2-amine 98%
5-Bromo-4-fluoropyridin-2-amine

Precaución: For research use only.

Formula: C₅H₄BrFN₂
MDL Number: MFCD16658678
CAS Number: 944401-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR