Bromo-PEG2-alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Bromoethoxy)ethanol

A210583-1G A210583-5G A210583-25G A210583-250MG
76825-878EA 13 USD
76825-878 77550-356 77568-034 77612-072
Bromo-PEG2-alcohol 95%
Bromo-PEG2-alcohol

Precaución: For research use only.

Formula: C₄H₉BrO₂
MDL Number: MFCD20621572
CAS Number: 57641-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR