2,5-Dichloro-4,6-dimethylnicotinic acid 97%

Supplier: Ambeed
Warning

A210325-100MG A210325-1G A210325-250MG
76825-860EA 42 USD
76825-860 76825-862 76825-864
2,5-Dichloro-4,6-dimethylnicotinic acid 97%
2,5-Dichloro-4,6-dimethylnicotinic acid

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MDL Number: MFCD02932725
CAS Number: 142266-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR