2-Bromo-N-methylaniline 95%

Supplier: Ambeed
Warning

A209629-10G A209629-1G A209629-250MG A209629-25G A209629-5G
76825-848EA 212.18 USD
76825-848 76825-850 76825-852 76825-854 77568-018
2-Bromo-N-methylaniline 95%
2-Bromo-N-methylaniline

Precaución: For research use only.

Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD06798064
CAS Number: 6832-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR