1-(2,2,2-Trifluoroethyl)piperazine hydrochloride 95%

Supplier: Ambeed
Warning

A209309-100MG A209309-250MG
76825-840EA 21.22 USD
76825-840 77683-595
1-(2,2,2-Trifluoroethyl)piperazine hydrochloride 95%
1-(2,2,2-Trifluoroethyl)piperazine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClF₃N₂
MW: 204.62 g/mol
MDL Number: MFCD10595946
CAS Number: 195447-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR