8-Chloro-6-iodoquinoline 95%

Supplier: Ambeed
Warning

A209306-100MG A209306-1G A209306-250MG
76825-834EA 45.62 USD
76825-834 76825-836 76825-838
8-Chloro-6-iodoquinoline 95%
8-Chloro-6-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClIN
MW: 289.50 g/mol
MDL Number: MFCD22376737
CAS Number: 111454-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR