2-Amino-4-methoxy-5-(3-morpholinopropoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A209287-1G A209287-250MG A209287-5G
76825-824EA 12 USD
76825-824 76825-826 76825-828
2-Amino-4-methoxy-5-(3-morpholinopropoxy)benzonitrile 97%
2-Amino-4-methoxy-5-(3-morpholinopropoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₅H₂₁N₃O₃
MDL Number: MFCD11110477
CAS Number: 675126-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR