2-Hydroxyisobutyric acid 97%

Supplier: Ambeed
Warning

A208292-100G A208292-10G A208292-25G A208292-500G A208292-5G A208292-1000G
76825-794EA 36 USD
76825-794 76825-796 76825-798 76825-800 76825-802 77885-508
2-Hydroxyisobutyric acid 97%
2-Hydroxyisobutyric acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MDL Number: MFCD00004459
CAS Number: 594-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR