Methyl 3-amino-2-(aminomethyl)propanoate dihydrochloride 95%

Supplier: Ambeed
Warning

A208180-100MG A208180-250MG
76825-790EA 69 USD
76825-790 76825-792
Methyl 3-amino-2-(aminomethyl)propanoate dihydrochloride 95%
Methyl 3-amino-2-(aminomethyl)propanoate dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂O₂
MW: 205.08 g/mol
MDL Number: MFCD21099537
CAS Number: 440644-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR