4-Bromo-2-chloroquinazoline 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloroquinazoline

A206798-100MG A206798-1G A206798-250MG A206798-5G
76825-764EA 102.91 USD
76825-764 76825-766 76825-768 76825-770
4-Bromo-2-chloroquinazoline 97%
4-Bromo-2-chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.4900 g/mol
MDL Number: MFCD11043856
CAS Number: 134517-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR