2-Amino-6-bromo-5-methylpyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-methylpyrazin-2-amine

A206375-100MG A206375-250MG A206375-1G
76825-756EA 45 USD
76825-756 76825-758 77885-495
2-Amino-6-bromo-5-methylpyrazine 97%
2-Amino-6-bromo-5-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MDL Number: MFCD17011819
CAS Number: 74290-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR