H-Chg-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Chg-OtBu.HCl

A206288-10G A206288-1G A206288-25G A206288-5G
76825-746EA 98.67 USD
76825-746 76825-748 76825-750 76825-752
H-Chg-OtBu·HCl 97%
H-Chg-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₂·HCl
MW: 249.8 g/mol
MDL Number: MFCD00270480
CAS Number: 213475-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR