2,3-Dibromo-6-methoxypyridine 97%

Supplier: Ambeed
Warning

A205933-100MG A205933-1G A205933-250MG A205933-5G
76825-738EA 13 USD
76825-738 76825-740 76825-742 77612-020
2,3-Dibromo-6-methoxypyridine 97%
2,3-Dibromo-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MDL Number: MFCD21648443
CAS Number: 909720-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR