5-Bromo-2,4-dimethyl-2H-indazole 95%

Supplier: Ambeed
Warning

A205855-1G A205855-250MG A205855-5G
76825-732EA 184.6 USD
76825-732 76825-734 76825-736
5-Bromo-2,4-dimethyl-2H-indazole 95%
5-Bromo-2,4-dimethyl-2H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
MDL Number: MFCD12028635
CAS Number: 1159511-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR