Ethyl 3-chloro-2-fluorobenzoate 97%

Supplier: Ambeed
Warning

A205725-1G A205725-250MG
76825-728EA 20.16 USD
76825-728 76825-730
Ethyl 3-chloro-2-fluorobenzoate 97%
Ethyl 3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
MDL Number: MFCD06204525
CAS Number: 773135-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR