4,7-Dibromo-1H-indole 98%

Supplier: Ambeed
Warning

A205675-100MG A205675-1G A205675-250MG
76825-720EA 28.65 USD
76825-720 76825-722 76825-724
4,7-Dibromo-1H-indole 98%
4,7-Dibromo-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
MDL Number: MFCD08272255
CAS Number: 126811-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR