(S)-Tetrahydrofuran-3-ylmethanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(Tetrahydrofuran-3-yl)methanol

A205473-1G A205473-250MG
76825-716EA 540 USD
76825-716 76825-718
(S)-Tetrahydrofuran-3-ylmethanol 97%
(S)-Tetrahydrofuran-3-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD17015961
CAS Number: 124391-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR