N-Methoxy-N-methylnicotinamide 95%

Supplier: Ambeed
Warning

A205201-5G A205201-1G A205201-25G
76825-714EA 33 USD
76825-714 77885-484 77885-485
N-Methoxy-N-methylnicotinamide 95%
N-Methoxy-N-methylnicotinamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD11847417
CAS Number: 95091-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR