3,5-Dibromo-2,4-dimethylpyridine 98%

Supplier: Ambeed
Warning

A205135-100MG A205135-1G A205135-5G
76825-708EA 55 USD
76825-708 76825-710 76825-712
3,5-Dibromo-2,4-dimethylpyridine 98%
3,5-Dibromo-2,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MDL Number: MFCD21337919
CAS Number: 29976-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR