2,3-Dihydrobenzo[b]furan-7-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzofuran-7-amine

A204961-100MG A204961-1G A204961-5G A204961-250MG
76825-702EA 38.2 USD
76825-702 76825-704 76825-706 77612-000
2,3-Dihydrobenzo[b]furan-7-ylamine 95%
2,3-Dihydrobenzo[b]furan-7-ylamine

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.163 g/mol
MDL Number: MFCD11109313
CAS Number: 13414-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR