5-Bromo-3-chloroisoquinoline 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A204950-100MG A204950-1G A204950-250MG A204950-5G
76825-692EA 19 USD
76825-692 76825-694 77593-334 77683-577
5-Bromo-3-chloroisoquinoline 95%, Ambeed.Inc
5-Bromo-3-chloroisoquinoline

Precaución: For research use only.

Formula: C₉H₅BrClN
MDL Number: MFCD11044700
CAS Number: 1029720-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR