2-(4-Chloro-2-hydroxyphenoxy)acetic acid 95%

Supplier: Ambeed
Warning

A204883-100MG A204883-10G A204883-1G A204883-250MG
76825-684EA 45.62 USD
76825-684 76825-686 76825-688 76825-690
2-(4-Chloro-2-hydroxyphenoxy)acetic acid 95%
2-(4-Chloro-2-hydroxyphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₄
MW: 202.59 g/mol
MDL Number: MFCD13192100
CAS Number: 7417-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR