(R)-4-(1-Aminoethyl)-N-(1H-pyrrolo[2,3-b]pyridin-4-yl)benzamide dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: Y-33075 2HCl

A204681-100MG A204681-250MG
76825-676EA 308.73 USD
76825-676 76825-678
(R)-4-(1-Aminoethyl)-N-(1H-pyrrolo[2,3-b]pyridin-4-yl)benzamide dihydrochloride 98+%
(R)-4-(1-Aminoethyl)-N-(1H-pyrrolo[2,3-b]pyridin-4-yl)benzamide dihydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₈Cl₂N₄O
MW: 353.24 g/mol
MDL Number: MFCD00945328
CAS Number: 173897-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR