2-Methylquinolin-7-ol 95%

Supplier: Ambeed
Warning

A204566-100MG A204566-1G A204566-250MG
76825-670EA 54.11 USD
76825-670 76825-672 76825-674
2-Methylquinolin-7-ol 95%
2-Methylquinolin-7-ol

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD18253749
CAS Number: 165112-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR