Methyl 2-(bromomethyl)-4-nitrobenzoate 95%

Supplier: Ambeed
Danger

A204520-100MG A204520-1G A204520-250MG A204520-5G
76825-664EA 30.77 USD
76825-664 76825-666 76825-668 77593-332
Methyl 2-(bromomethyl)-4-nitrobenzoate 95%
Methyl 2-(bromomethyl)-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.068 g/mol
MDL Number: MFCD11505948
CAS Number: 133446-99-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR