1,3-Dichloro-6-fluoroisoquinoline 95%

Supplier: Ambeed
Warning

A204481-1G A204481-250MG
76825-660EA 23 USD
76825-660 76825-662
1,3-Dichloro-6-fluoroisoquinoline 95%
1,3-Dichloro-6-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₄Cl₂FN
MDL Number: MFCD11656163
CAS Number: 1041423-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR