tert-Butyl 4-((tert-butyldiphenylsilyl)oxy)cyclohexanecarboxylate 95%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A204348-10G
A204348-1G
76825-658EA
1118
USD
76825-658
77683-575
tert-Butyl 4-((tert-butyldiphenylsilyl)oxy)cyclohexanecarboxylate 95%, Ambeed.Inc
tert-Butyl 4-((tert-butyldiphenylsilyl)oxy)cyclohexanecarboxylate
Precaución: For research use only.
Formula:
C₂₇H₃₈O₃Si MW: 438.67 g/mol |
MDL Number:
MFCD17392810 CAS Number: 1010086-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...