(2,5-Dibromophenyl)methanol 98%

Supplier: Ambeed
Warning

A204185-1G A204185-250MG A204185-10G A204185-5G
76825-652EA 13 USD
76825-652 76825-654 77567-954 77885-478
(2,5-Dibromophenyl)methanol 98%
(2,5-Dibromophenyl)methanol

Precaución: For research use only.

Formula: C₇H₆Br₂O
MDL Number: MFCD12031827
CAS Number: 147034-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR