5-Bromo-4-fluoro-2-nitroaniline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4-bromo-5-fluoronitrobenzene
A204176-1G
A204176-5G
A204176-25G
76825-648EA
16.98
USD
76825-648
76825-650
77683-573
5-Bromo-4-fluoro-2-nitroaniline 97%
5-Bromo-4-fluoro-2-nitroaniline
Precaución: For research use only.
Formula:
C₆H₄BrFN₂O₂ MW: 235.011 g/mol |
MDL Number:
MFCD21332915 CAS Number: 1052686-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...