3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A203982-1G A203982-250MG A203982-5G
76825-642EA 107.16 USD
76825-642 76825-644 76825-646
3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester 98%
3-Chloro-4-(ethoxycarbonyl)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BClO₄
MW: 310.581 g/mol
MDL Number: MFCD06659929
CAS Number: 474709-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR