2-Bromo-4-methyl-5-nitroaniline 95%

Supplier: Ambeed
Warning

A203910-10G A203910-1G A203910-250MG A203910-5G
76825-634EA 135 USD
76825-634 76825-636 76825-638 76825-640
2-Bromo-4-methyl-5-nitroaniline 95%
2-Bromo-4-methyl-5-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD01239994
CAS Number: 102169-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR