STAT3 Inhibitor III, WP1066 ≥98%

Supplier: Ambeed
Warning

Synonyms: WP1066, (S,E)-3-(6-Bromopyridin-2-yl)-2-cyano-N-(1-phenylethyl)acrylamide, State3 Inhibitor III

A203794-10MG A203794-1G A203794-1MG A203794-100MG
76825-628EA 9.55 USD
76825-628 76825-630 76825-632 77683-571
STAT3 Inhibitor III, WP1066 ≥98%
STAT3 Inhibitor III, WP1066

Precaución: For research use only.

Formula: C₁₇H₁₄BrN₃O
MW: 356.22 g/mol
MDL Number: MFCD12912450
CAS Number: 857064-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR