AN-2728 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-((1-hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-6-yl)oxy)benzonitrile, 4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)benzonitrile, Crisaborole

A203325-10MG A203325-50MG A203325-100MG A203325-1G A203325-1MG A203325-25G
76825-620EA 5.31 USD
76825-620 76825-622 77611-972 77611-974 77683-567 77683-568
AN-2728 98%, Ambeed.Inc
AN-2728

Precaución: For research use only.

Formula: C₁₄H₁₀BNO₃
MW: 251.04 g/mol
MDL Number: MFCD17169940
CAS Number: 906673-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR