2-Amino-4-bromo-6-methylphenol 95%

Supplier: Ambeed
Warning

A203085-1G A203085-250MG
76825-610EA 127.31 USD
76825-610 76825-612
2-Amino-4-bromo-6-methylphenol 95%
2-Amino-4-bromo-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD18294016
CAS Number: 343269-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR