Ethyl-5-methyl-5-hexenoate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-methylhex-5-enoate

A201844-1G A201844-250MG
76825-586EA 145.35 USD
76825-586 76825-588
Ethyl-5-methyl-5-hexenoate 95%
Ethyl-5-methyl-5-hexenoate

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
MDL Number: MFCD00671855
CAS Number: 39495-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR