2-Chloro-6-iodobenzaldehyde 95%

Supplier: Ambeed
Warning

A201275-100MG A201275-250MG
76825-576EA 40.32 USD
76825-576 76825-578
2-Chloro-6-iodobenzaldehyde 95%
2-Chloro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.464 g/mol
MDL Number: MFCD18393320
CAS Number: 51738-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR