Ethyl 4-chloro-3-fluorobenzoate 98%

Supplier: Ambeed
Warning

A200740-100G A200740-10G A200740-25G
76825-568EA 123 USD
76825-568 76825-570 76825-572
Ethyl 4-chloro-3-fluorobenzoate 98%
Ethyl 4-chloro-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MDL Number: MFCD06204445
CAS Number: 203573-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR