5-Chloro-7H-pyrrolo[2,3-d]pyrimidine 95%

Supplier: Ambeed
Warning

A200561-100MG A200561-1G A200561-250MG A200561-5G
76825-552EA 15 USD
76825-552 76825-554 76825-556 76825-558
5-Chloro-7H-pyrrolo[2,3-d]pyrimidine 95%
5-Chloro-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MDL Number: MFCD12031334
CAS Number: 1041864-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR