3-Ethylbenzene-1,2-diol 95%

Supplier: Ambeed
Warning

A199658-1G A199658-250MG
76825-524EA 110.34 USD
76825-524 76825-526
3-Ethylbenzene-1,2-diol 95%
3-Ethylbenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.16 g/mol
MDL Number: MFCD21604516
CAS Number: 933-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR