(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-Pyroglutamic Acid, (S)-(-)-2-Pyrrolidone-5-carboxylic acid, H-Pyr-OH, (S)-5-Oxopyrrolidine-2-carboxylic acid

A199648-1000G A199648-100G A199648-25G A199648-500G
76825-516EA 32.89 USD
76825-516 76825-518 76825-520 76825-522
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98%
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₇NO₃
MW: 129.1 g/mol
MDL Number: MFCD00005272
CAS Number: 98-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR