3-(Piperidin-4-yl)indolin-2-one 95%

Supplier: Ambeed
Warning

A199207-100MG A199207-1G A199207-250MG
76825-504EA 227.04 USD
76825-504 76825-506 76825-508
3-(Piperidin-4-yl)indolin-2-one 95%
3-(Piperidin-4-yl)indolin-2-one

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O
MW: 216.27 g/mol
MDL Number: MFCD18801077
CAS Number: 72831-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR