2-(4-Fluorophenyl)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A198909-100MG A198909-1G A198909-250MG A198909-5G
76825-490EA 25 USD
76825-490 76825-492 76825-494 77885-463
2-(4-Fluorophenyl)cyclopropanecarboxylic acid 95%
2-(4-Fluorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
MDL Number: MFCD06804481
CAS Number: 879324-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR