Methyl 4-bromo-2-(trifluoromethyl)benzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2-(trifluoromethyl)benzoic acid methyl ester
A198694-100G
A198694-1G
A198694-25G
A198694-5G
76825-482EA
704.44
USD
76825-482
76825-484
76825-486
76825-488
Methyl 4-bromo-2-(trifluoromethyl)benzoate 98%
Methyl 4-bromo-2-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₉H₆BrF₃O₂ MW: 283.042 g/mol |
MDL Number:
MFCD12761599 CAS Number: 957207-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...