2-(5-Bromo-2-methylphenyl)acetic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A198233-1G A198233-5G A198233-100MG A198233-250MG
76825-468EA 70 USD
76825-468 77683-544 77885-459 77885-460
2-(5-Bromo-2-methylphenyl)acetic acid 97%, Ambeed.Inc
2-(5-Bromo-2-methylphenyl)acetic acid

Precaución: For research use only.

Formula: C₉H₉BrO₂
MDL Number: MFCD11840270
CAS Number: 854646-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR