3-Bromo-7-methoxy-4H-chromen-4-one 95%

Supplier: Ambeed
Warning

A197327-100MG A197327-250MG
76825-452EA 60.48 USD
76825-452 76825-454
3-Bromo-7-methoxy-4H-chromen-4-one 95%
3-Bromo-7-methoxy-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₃
MW: 255.06 g/mol
MDL Number: MFCD20528125
CAS Number: 73220-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR